Learn More
Sino Biological Anti-CD39/ENTPD1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 109343T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse CD39, and purified by antigen affinity chromatography.
Catalog No. 50-255-5048