Learn More
Sino Biological Anti-CD39/ENTPD1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50398RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD39 (Catalog#50398-M08B; P55772; Thr38-Ile478). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3284