Learn More
American Research Products Inc Mouse eosinophil cationic prot
Catalog No. 5000329961
Encompass
Mouse eosinophil cationic protein ECP ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29961 Supplier American Research Products Inc Supplier No. CSBE11799M
Mouse eosinophil cationic protein ECP ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich