Learn More
Sino Biological Mouse EpCAM MAb 100ul (Sino Biological)
Supplier:  Sino Biological 50591R065100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EpCAM / TROP-1 / TACSTD1 (rM EpCAM / TROP-1 / TACSTD1; Catalog#50591-M08H; NP_032558.2; Met1-Thr266).
Catalog No. 50-237-3055