Learn More
Sino Biological Recombinant Anti-EpCAM/TROP1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50591R052100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EpCAM / TROP-1 / TACSTD1 (rM EpCAM / TROP-1 / TACSTD1; Catalog#50591-M08H; NP_032558.2; Met1-Thr266).
Catalog No. 50-240-8484