Learn More
BIOHIPPO Mouse EPCAM Microsample
SDP
Catalog No. 5000313751
Encompass
Mouse EPCAM Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of EPCAM in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 15 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse EPCAM Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse EPCAM Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse EPCAM biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13751 Supplier BIOHIPPO Supplier No. BHE15212378
Mouse EPCAM Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of EPCAM in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 15 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse EPCAM Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse EPCAM Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse EPCAM biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color