Learn More
Sino Biological Mouse EpCAM/TROP1 Matched ELISA Antibody Pair Set 15 Plates
Supplier:  Sino Biological SEK5059115
Encompass_Preferred
The Mouse EpCAM / TROP-1 / TACSTD1 ELISA Pair Set is for the quantitative determination of Mouse EpCAM / TROP-1 / TACSTD1.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for EpCAM / TROP-1 / TACSTD1 coated on a 96-well plate. Standards and samples are added to the wells, and any EpCAM / TROP-1 / TACSTD1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-EpCAM / TROP-1 / TACSTD1 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of EpCAM / TROP-1 / TACSTD1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450
Catalog No. 50-007-85604