Learn More
Biomatik Corporation Mouse Ependymin Related Protein 1 (EPDR1) ELISA Kit
Supplier:  Biomatik Corporation EKU12261
Encompass_Preferred
Mouse Ependymin Related Protein 1 (EPDR1) ELISA Kit for detecting EPDR1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99M71. Gene ID: 105298. Target Synonyms: MERP1; UCC1; EPDR; Upregulated In Colorectal Cancer Gene-1; Mammalian ependymin-related protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7992