Learn More
Sino Biological Recombinant Anti-EphA2 Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 50586R301F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EphA2 / Eph Receptor A2 (rM EphA2 / Eph Receptor A2; Catalog#50586-M08H; NP_034269.2; Met1-Asn535) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-255-3199