Learn More
Sino Biological Mouse EphA2 MAb 100TS (Sino Biological)
Supplier:  Sino Biological 50586R101A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EphA2/Eph Receptor A2 (rM EphA2/Eph Receptor A2; Catalog#50586-M08H; NP_034269.2; Met1-Asn535) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-237-2712