Learn More
Sino Biological Recombinant Mouse EphA2 Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 50586M49HB20
Encompass_Preferred
A DNA sequence encoding the Mouse EphA2 (NP_034269.2) (Met1-Asn535) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-2901