Learn More
Sino Biological Mouse EphA6 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 50630R033100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EphA6 / EHK2 (rM EphA6 / EHK2; Catalog#50630-M08H; NP_031964.2; Met 1-Gln 546).
Catalog No. 50-237-1480