Learn More
Sino Biological Anti-EphA6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50630RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse EphA6 (rM EphA6; Catalog#50630-M08H; NP_031964.2; Met 1-Gln 546). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-7727