Learn More
Sino Biological Anti-EphA7 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50587RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse EphA7 / Eph Receptor A7 (Catalog#50587-M08H; Q61772-1; Met1-Ile556). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0806