Learn More
Sino Biological Mouse EphB4 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 50582R039100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EphB4 (rM EphB4; Catalog#50582-M08H; P54761-1; Met 1-Ala 539).
Catalog No. 50-237-3501