Learn More
Sino Biological Anti-EphB4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50582RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse EphB4 (rM EphB4; Catalog#50582-M08H; P54761-1; Met 1-Ala 539). EphB4 specific IgG was purified by Mouse EphB4 affinity chromatography.
Catalog No. 50-255-3206