Learn More
Sino Biological Anti-EphB6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51123T08100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse EphB6 / Eph Receptor B6 (rM EphB6 / Eph Receptor B6; Catalog#51123-M08H; O08644-1; Met1-Ser587). EphB6 / Eph Receptor B6 specific IgG was purified by Mouse EphB6 / Eph Receptor B6 affinity chromatography.
Catalog No. 50-255-2997