Learn More
American Research Products Inc Mouse Epidermal growth factor
Supplier:  American Research Products Inc CSBEL007476MO
Encompass
Mouse Epidermal growth factor-like protein 7 (EGFL7) ELISA kit from CUSABIO Detection Range 7 8 ng/mL-500 ng/mL Sensitivity 1 95 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27923