Learn More
American Research Products Inc Mouse Epidermal growth factor
Catalog No. 5000328167
Encompass
Mouse Epidermal growth factor-like protein 6 (EGFL6) ELISA kit from CUSABIO Detection Range 93 75 pg/mL-6000 pg/mL Sensitivity 23 43 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28167 Supplier American Research Products Inc Supplier No. CSBEL007475MO
Mouse Epidermal growth factor-like protein 6 (EGFL6) ELISA kit from CUSABIO Detection Range 93 75 pg/mL-6000 pg/mL Sensitivity 23 43 pg/mL Sample serum plasma tissue homogenates Method Sandwich