Learn More
American Research Products Inc Mouse Epinephrine Adrenaline
Supplier:  American Research Products Inc CSBE08679M
Encompass
Mouse Epinephrine/Adrenaline EPI ELISA Kit from CUSABIO Detection Range 0 78 ng/ml-50 ng/ml Sensitivity 0 2 ng/ml Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29720