Learn More
American Research Products Inc Mouse Epithelial neutrophil ac
Catalog No. 5000328827
Encompass
Mouse Epithelial neutrophil actiVating peptide 78 ENA-78 ELISA Kit from CUSABIO Detection Range 62 5 pg/ml - 4000 pg/ml Sensitivity 15 6 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28827 Supplier American Research Products Inc Supplier No. CSBE08180M
Mouse Epithelial neutrophil actiVating peptide 78 ENA-78 ELISA Kit from CUSABIO Detection Range 62 5 pg/ml - 4000 pg/ml Sensitivity 15 6 pg/ml Sample serum plasma tissue homogenates Method Sandwich