Learn More
Kingfisher Biotech Inc Rabbit EPO ELISA - Less than or equal to 10 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1588M004
Encompass_Preferred
The Rabbit Erythropoietin (EPO) ELISA contains capture antibody, standard, and detection antibody for development of a Rabbit Erythropoietin ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. Alternate names include EPO, EP, MVCD2, erythropoietin, Erythropoietin, ECYT5, DBAL. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5538