Learn More
BIOHIPPO Mouse EPO Microsample
SDP
Catalog No. 5000314107
Encompass
Mouse EPO Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of EPO in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 6 25-400 pg/mL with a sensitivity of 2 84 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse EPO Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse EPO Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse EPO biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14107 Supplier BIOHIPPO Supplier No. BHE15212342
Mouse EPO Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of EPO in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 6 25-400 pg/mL with a sensitivity of 2 84 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse EPO Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse EPO Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse EPO biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color