Learn More
Sino Biological Anti-Erythropoietin/EPO Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51099T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Erythropoietin (rM Erythropoietin; Catalog#51099-MNAH; NP_031968.1; Met1-Arg192). Erythropoietin specific IgG was purified by Mouse Erythropoietin affinity chromatography.
Catalog No. 50-240-8258