Learn More
Sino Biological Anti-Erythropoietin R/EPOR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50031T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse EPOR / Erythropoietin Receptor (rM EPOR / Erythropoietin Receptor; Catalog#50031-M08H; NP_034279.3; Met1-Pro249). EPOR / Erythropoietin Receptor specific IgG was purified by Mouse EPOR / Erythropoietin Receptor affinity chromatography.
Catalog No. 50-255-3544