Learn More
Sino Biological Recombinant Anti-Her2/ERBB2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50714R007100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Her2/ERBB2 (rM Her2/ERBB2; Catalog#50714-M08H; P70424; Met1-Thr653).
Catalog No. 50-240-8239