Learn More
Sino Biological Recombinant Anti-Her2/ERBB2 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 50714R001P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse HER2 / ErbB2 / CD340 (rM HER2 / ErbB2 / CD340; Catalog#50714-M08H; P70424; Met1-Thr653) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-240-8234