Learn More
Sino Biological Mouse ERBB3 ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502373608
Encompass_Preferred
Each vial contains 57 ng of recombinant Mouse ERBB3/HER3. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2500 pg/mL is recommended.
Catalog No. 50-237-3608 Supplier Sino Biological Supplier No. SEK5100315
Each vial contains 57 ng of recombinant Mouse ERBB3/HER3. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2500 pg/mL is recommended.