Learn More
Sino Biological Recombinant Anti-HER3/ERBB3 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 51003R028100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse HER3 / ErbB3 (rM HER3 / ErbB3; Catalog#51003-M08H; Q61526; Met1-His641).
Catalog No. 50-241-0040