Learn More
Sino Biological Mouse ERBB3 Protein 20ug (Sino Biological)
Supplier:  Sino Biological 81069R08H100
Encompass_Preferred
A DNA sequence encoding the Mouse ERBB3 (Q61526) (Met1-His641) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-4366