Learn More
Biomatik Corporation Mouse Erythropoietin (EPO) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11100
Encompass_Preferred
Mouse Erythropoietin (EPO) Mini sample ELISA Kit for detecting EPO in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.25-400pg/ml. Sensitivity: 2.39pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07321. Gene ID: 13856. Target Synonyms: EP; Epoetin; Erythropoetin; Hematopoietin; Hemopoietin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8225