Learn More
Sino Biological Anti-ER alpha Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106132T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse ER alpha/ESR1, and purified by antigen affinity chromatography.
Catalog No. 50-240-8981