Learn More
Sino Biological Anti-ESRRB Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107441T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse ESRRB, and purified by antigen affinity chromatography.
Catalog No. 50-241-0929