Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse Estradiol Rapid ELISA Kit

Catalog No. EELR013
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EELR013 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EELR013 Supplier Invitrogen™ Supplier No. EELR013
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Mouse Estradiol (E2) ELISA quantitates E2 in Serum, Plasma, Urine, Saliva. Principle of the method The Mouse E2 solid phase competitive ELISA (enzyme linked immunosorbent assay) is designed to measure the amount of the target based on competitive binding. A target specific antibody has been pre coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells, where Mouse E2 in the samples competes with the pre coated Mouse E2 for binding sites on the Horseradish Peroxidase (HRP) linked detection antibody. Excess conjugate and unbound sample or standard are washed away. A substrate solution is added that reacts with the enzyme antibody target complex to produce a measurable signal. The intensity of this signal, measured as optical density (OD) at a wavelength of 450±2 nm, is inversely proportional to the concentration of target present in the original specimen. The concentration of Mouse E2 in the samples is then determined by comparing the OD of the samples to the standard curve. Achieve fast and reliable results in just 90 minutes with our Rapid ELISA kit, featuring a streamlined protocol that requires only one wash step. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation. For Research Use Only. Not for use in diagnostic procedures. Not for resale wit...

Estradiol (17 beta-estradiol) is a sex hormone. Estradiol is the predominant sex hormone present in females. It is also present in males, being produced as an active metabolic product of testosterone. It represents the major estrogen in humans. Estradiol has not only a critical impact on reproductive and sexual functioning, but also affects other organs including the bones.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 3.13-200 pg/mL
Assay Sensitivity 1.88 pg/mL
Conjugate HRP
Immunoassay Kit Format Coated ELISA Kit
Product Type ELISA
Sample Type Plasma, Saliva, Serum, Urine
Target Species Mouse
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 17-Beta-Estradiol; 17-beta-HSD17-Beta-Estradiol; 17-beta-HSD;
Interassay CV <10%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, HRP Conjugate, Standard & Sample Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer, Standard, HRP Conjugate, Standard & Sample Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Saliva 50 μL, Serum 50 μL, Urine 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Estradiol
Test Time 15 min
Total Assay Time 1 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.