Learn More
Abcam Mouse Estrogen ELISA Kit, 96TEST
Supplier:  Abcam AB28529196TEST
Encompass_Preferred
Mouse Estrogen ELISA Kit is a Competitive ELISA used to quantify Mouse Estrogen with a sensitivity of 9.375 pg/ml - Colorimetric ELISA kit - 450 nm readout. Works on any standard plate reader - Wide dynamic range – 15.625 - 1000 pg/ml
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5472