Learn More
American Research Products Inc Mouse EXtracellular superoXide
Catalog No. 5000328244
Encompass
Mouse EXtracellular superoXide dismutase [Cu-Zn] (SOD3) ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28244 Supplier American Research Products Inc Supplier No. CSBEL022399MO
Mouse EXtracellular superoXide dismutase [Cu-Zn] (SOD3) ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich