Learn More
American Research Products Inc Mouse F boX only protein 32 F
Catalog No. 5000328169
Encompass
Mouse F-boX only protein 32 (FBXO32) ELISA kit from CUSABIO Detection Range 37 5 pg/mL-2400 pg/mL Sensitivity 9 375 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28169 Supplier American Research Products Inc Supplier No. CSBEL008498MO
Mouse F-boX only protein 32 (FBXO32) ELISA kit from CUSABIO Detection Range 37 5 pg/mL-2400 pg/mL Sensitivity 9 375 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich