Learn More
Sino Biological Anti-Coagulation Factor II Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50344T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Coagulation Factor II / F2 (rM Coagulation Factor II / F2; Catalog#50344-M08H; NP_034298.1; Met1-Gly618). Coagulation Factor II / F2 specific IgG was purified by Mouse Coagulation Factor II / F2 affinity chromatography.
Catalog No. 50-255-3301