Learn More
Sino Biological Recombinant Anti-Tissue Factor Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50413R034100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Coagulation Factor III / Tissue Factor / CD142 / F3 (rM Coagulation Factor III / Tissue Factor / CD142 / F3; Catalog#50413-M08H; NP_034301.3; Met 1-Glu 251).
Catalog No. 50-241-0510