Learn More
Sino Biological Anti-Tissue Factor Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50413RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse F3 / CD142 (rM F3; Catalog#50413-M08H; NP_034301.3; Met 1-Glu 251). F3 / CD142 specific IgG was purified by mouse F3 / CD142 affinity chromatography.
Catalog No. 50-241-2487