Learn More
Sino Biological Mouse F7 ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502370515
Encompass_Preferred
Each vial contains 40 ng of recombinant Mouse Factor VII/F7. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 2000 pg/mL is recommended.
Catalog No. 50-237-0515 Supplier Sino Biological Supplier No. SEK5003415
Each vial contains 40 ng of recombinant Mouse Factor VII/F7. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 2000 pg/mL is recommended.