Learn More
Vector Laboratories Mouse F7 / Factor VII (Sandwich ELISA) ELISA Kit
SDP
Supplier:  Vector Laboratories LSF114091
Encompass
96-well enzyme-linked immunosorbent assay (ELISA) for the Quantitative detection of Mouse F7 / Factor VII in samples of Plasma and Serum. It is based upon a Sandwich assay principle and can be used to detect levels of F7 / Factor VII as low as 7.8 nanograms per millilter.
Catalog No. NC1847075