Learn More
Sino Biological Recombinant Anti-Factor VII Antibody (HRP), Rabbit Monoclonal
Supplier:  Sino Biological 50034R121H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse FVII (rM FVII; Catalog#50034-M08C; NP_034302.2; Met 1-Leu 446), and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-255-3539