Learn More
Sino Biological Recombinant Anti-Factor VII Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50034R003100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Coagulation Factor VII (rM Coagulation Factor VII; Catalog#50034-M08C; NP_034302.2; Met1-Leu446).
Catalog No. 50-255-3543