Learn More
BIOHIPPO Mouse F7 Microsample
SDP
Catalog No. 5000314134
Encompass
Mouse F7 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of F7 in Plasma It provides a detection range of 15 63-1000 ng/mL with a sensitivity of 0 3 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse F7 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse F7 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse F7 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14134 Supplier BIOHIPPO Supplier No. BHE15212553
Mouse F7 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of F7 in Plasma It provides a detection range of 15 63-1000 ng/mL with a sensitivity of 0 3 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse F7 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse F7 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse F7 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color