Learn More
Sino Biological Anti-Factor VII Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50034RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, Stable recombinant Mouse FVII / Factor VII (rM FVII; Catalog#50034-M08C; NP_034302.2; Met 1-Leu 446). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3537