Learn More
BIOHIPPO Mouse F8 Microsample
SDP
Catalog No. 5000314767
Encompass
Mouse F8 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of F8 in plasma It provides a detection range of 0 32-20 IU/mL with a sensitivity of 0 081 IU/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse F8 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse F8 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse F8 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14767 Supplier BIOHIPPO Supplier No. BHE15212588
Mouse F8 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of F8 in plasma It provides a detection range of 0 32-20 IU/mL with a sensitivity of 0 081 IU/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse F8 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse F8 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse F8 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color