Learn More
Sino Biological Mouse F9 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 50362R00320
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Coagulation Factor IX / FIX / F9 (Catalog#50362-M08H; P16294; Met1-Thr471).
Catalog No. 50-237-3098