Learn More
Sino Biological Mouse F9 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50362T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Coagulation Factor IX / FIX / F9 (Catalog#50362-M08H; P16294; Met1-Thr471). Coagulation Factor IX / FIX / F9 specific IgG was purified by Mouse Coagulation Factor IX / FIX / F9 affinity chromatography.
Catalog No. 50-237-2858