Learn More
Biomatik Corporation Mouse Factor Related Apoptosis Ligand (FASL) CLIA Kit
Supplier:  Biomatik Corporation EKU08780
Encompass_Preferred
Mouse Factor Related Apoptosis Ligand (FASL) CLIA Kit for detecting FASL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 41.2-30,000pg/ml. Sensitivity: 15.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q544E9. Gene ID: 14103. Target Synonyms: CD178; CD95L; CD95-L; FASLG; FASLG; APT1LG1; APT1-LG1; TNFSF6; Fas Antigen Ligand; TNF Superfamily Member 6; Tumor Necrosis Factor (ligand) Superfamily,Member 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7540